Learn More
Strem, An Ascensus Company CAS# 10026-12-7. 25g. Niobium(V) chloride (99.99%-Nb) (20-200ppm Ta) PURATREM. MFCD00011127
SDP
Catalog No. 5090111647
Encompass
CAS# 10026-12-7. 25g. Niobium(V) chloride (99.99%-Nb) (20-200ppm Ta) PURATREM. MFCD00011127. Molecular Weight: 270.17. Molecular Formula: NbCl5. Color/form: yellow xtl. Strem# 41-1250. http://www.strem.com/catalog/v/41-1250/
Catalog No. 50-901-11647 Supplier Strem, An Ascensus Company Supplier No. 41125025G
CAS# 10026-12-7. 25g. Niobium(V) chloride (99.99%-Nb) (20-200ppm Ta) PURATREM. MFCD00011127. Molecular Weight: 270.17. Molecular Formula: NbCl5. Color/form: yellow xtl. Strem# 41-1250. http://www.strem.com/catalog/v/41-1250/