Niobium(V) chloride, 99% (metals basis), Alfa Aesar™

For Use in Preparation of Pure Niobium, and As An Intermediate

Manufacturer: Alfa Aesar 5110809

Quantity 10g

 View more versions of this product

Catalog No. AA5110809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10026-12-7
    Physical Form 15mm and down Lumps, Packaged Under Argon
    Assay Percent Range 99% (metals basis)
    Molecular Formula NbCl5
    Formula Weight 270.2
    MDL Number MFCD00011127
    Synonym Niobium pentachloride
    Chemical Name or Material Niobium(V) chloride

    Specifications

    Physical Form 15mm and down Lumps, Packaged Under Argon
    Formula Weight 270.2
    Quantity 10g
    Color Yellow
    DOT Information DOT Class: 8, Packaging Group: II
    Density 2.74g/cm3 at 20°C
    EINECS Number 233-059-8
    Odor Acidic
    Assay Percent Range 99% (metals basis)
    Molecular Formula NbCl5
    MDL Number MFCD00011127
    CAS 10026-12-7
    Synonym Niobium pentachloride
    Health Hazard 1 H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.
    Chemical Name or Material Niobium(V) chloride

    H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit