Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Niobium(V) ethoxide

Catalog No. 3392025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-920-25G 5 g
33-920-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-920-25G Supplier Sigma Aldrich Supplier No. 3392025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Niobium(V) ethoxide

Specifications

Boiling Point 142°C/0.1, mmHg
Melting Point 5°C to 6°C
CAS 3236-82-6
Quantity 5 g
Molecular Formula C10H25NbO5
Linear Formula Nb(OCH2CH3)5
MDL Number MFCD00015122
Synonym NbOEt; Niobium ethanolate; Niobium ethylate; Niobium pentaethoxide; Pentaethoxyniobium
Molecular Weight (g/mol) 318.21
Percent Purity 99.95% trace metals basis
Refractive Index n20/D 1.516 (literature)
Density 1.268 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.