Learn More
Strem, An Ascensus Company CAS# 1313-96-8. 250g. Niobium(V) oxide (99.5%-Nb). MFCD00011128
SDP
Catalog No. 5090115302
Encompass
CAS# 1313-96-8. 250g. Niobium(V) oxide (99.5%-Nb). MFCD00011128. Molecular Weight: 265.82. Molecular Formula: Nb2O5. Color/form: white pwdr. Strem# 93-4111. http://www.strem.com/catalog/v/93-4111/
Catalog No. 50-901-15302 Supplier Strem, An Ascensus Company Supplier No. 934111250G
CAS# 1313-96-8. 250g. Niobium(V) oxide (99.5%-Nb). MFCD00011128. Molecular Weight: 265.82. Molecular Formula: Nb2O5. Color/form: white pwdr. Strem# 93-4111. http://www.strem.com/catalog/v/93-4111/