Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ NIOSH Respirator Cartridges

Catalog No. 19088340 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask
Product Type:
Cartridge
Color:
Black/Pink
Filter Type:
P100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type Color Filter Type
19-088-340 X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask Cartridge Black/Pink P100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-340 Supplier Dräger™ Supplier No. 6738034
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Cartridge Type Organic Vapor Cartridge
Certifications/Compliance NIOSH
Color Black/Pink
Filter Type P100
Product Type Cartridge
For Use With (Application) To purify air
For Use With (Equipment) X-plore™ 3300 and 3500 half mask, x-plore™ 5500 full face mask
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.