Learn More
Aviva Systems Biology Corp NIP7 Antibody - C-terminal region (ARP40833_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40833_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NIP7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NIP7. Alternate names: KD93, CGI-37, HSPC031ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57635