Learn More
Aviva Systems Biology Corp NIP7 Antibody - middle region (ARP40832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NIP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NIP7. Alternate names: KD93, CGI-37, HSPC031ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57634