Learn More
Aviva Systems Biology Corp NIPA1 Antibody - C-terminal region (ARP64519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NIPA1. It was validated on Western Blot. . Alternate names: FSP3, MGC102724, MGC35570, SPG6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-281