Learn More
ACROBiosystems ACROBiosystems Nipah virus Fusion glycoprotein, His Tag
Catalog No. 502107190
Encompass_Preferred
Recombinant Protein;50UG;Nipah virus Fusion glycoprotein, His Tag (FUN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Accession # Q9IH63-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.9 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Fusion Protein Antibody, Human IgG1 (5B3) at 1 μg/mL (100 μL/well) can bind Nipah virus Fusion glycoprotein, His Tag (Cat. No. FUN-N52H3) with a linear range of 1-2 ng/mL (QC tested).
Catalog No. 50-210-7190 Supplier ACROBiosystems Supplier No. FUNN52H350UG
Recombinant Protein;50UG;Nipah virus Fusion glycoprotein, His Tag (FUN-N52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Accession # Q9IH63-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.9 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Fusion Protein Antibody, Human IgG1 (5B3) at 1 μg/mL (100 μL/well) can bind Nipah virus Fusion glycoprotein, His Tag (Cat. No. FUN-N52H3) with a linear range of 1-2 ng/mL (QC tested).