Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nipecotamide, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
4138-26-5
C6H12N2O
128.17
MFCD00005993
BVOCPVIXARZNQN-UHFFFAOYSA-N
nipecotamide,3-piperidinecarboxamide,nipecotic acid amide,hexahydronicotinamide,piperidine-3-carboxylic acid amide,racemic nipecotamide,pubchem11797,acmc-20lak2,3-piperidine carboxamide
92980
CHEBI:60116
piperidine-3-carboxamide
C1CC(CNC1)C(=O)N
4138-26-5
C6H12N2O
128.17
MFCD00005993
BVOCPVIXARZNQN-UHFFFAOYSA-N
nipecotamide,3-piperidinecarboxamide,nipecotic acid amide,hexahydronicotinamide,piperidine-3-carboxylic acid amide,racemic nipecotamide,pubchem11797,acmc-20lak2,3-piperidine carboxamide
92980
CHEBI:60116
piperidine-3-carboxamide
C1CC(CNC1)C(=O)N
Melting Point 102°C to 108°C
Color Yellow
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 128.17
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material Nipecotamide, 95%
Show More Show Less