Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nipecotamide 98.0+%, TCI America™
SDP

Catalog No. N00975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00975G 5 g
N009725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N00975G Supplier TCI America Supplier No. N00975G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4138-26-5 | C6H12N2O | 128.175 g/mol

Chemical Identifiers

CAS 4138-26-5
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00005993
InChI Key BVOCPVIXARZNQN-UHFFFAOYSA-N
Synonym nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide
PubChem CID 92980
ChEBI CHEBI:60116
IUPAC Name piperidine-3-carboxamide
SMILES C1CC(CNC1)C(=O)N

Specifications

Melting Point 111°C
Quantity 5 g
Formula Weight 128.18
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Nipecotamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.