Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(±)-Nipecotic acid, Tocris Bioscience™
Description
GABA uptake inhibitor. IC50 values are 8, 38, 106 and 2370μM for hGAT-1, rGAT-2, hGAT-3 and hBGT-1 respectively.
Specifications
Specifications
| Quantity | 100 mg |
| Formula Weight | 129.16 |
| Percent Purity | >98% |
| Chemical Name or Material | (±)-Nipecotic acid |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?