Learn More
World Precision Instrument Cuticle Nipper, 11cm
SDP
Catalog No. 50822372
Encompass
Ring Handled Cuticle Nippers are a handy instrument to have in a life science laboratory. While they are designed for trimming excess cuticles, hangnails, and dead skin around the nails, the unique shape makes them versatile. When working with plants or animals, you can precisely trim sections of tissue in tight spots. You can clip the ends of sutures, cleanly prune young plants, trim away exoskeleton component during microdissection, or collect tissue samples. The ring handles on our cuticle nippers give you excellent control. The angled blades allow you to get clean cuts in tight spaces, and they give you a clear view of the cutting area. These surgical instruments are manufactured from surgical grade stainless steel or premium German steel, both of which are suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.
Catalog No. 50-822-372 Supplier World Precision Instrument Supplier No. 500137
Ring Handled Cuticle Nippers are a handy instrument to have in a life science laboratory. While they are designed for trimming excess cuticles, hangnails, and dead skin around the nails, the unique shape makes them versatile. When working with plants or animals, you can precisely trim sections of tissue in tight spots. You can clip the ends of sutures, cleanly prune young plants, trim away exoskeleton component during microdissection, or collect tissue samples. The ring handles on our cuticle nippers give you excellent control. The angled blades allow you to get clean cuts in tight spaces, and they give you a clear view of the cutting area. These surgical instruments are manufactured from surgical grade stainless steel or premium German steel, both of which are suitable for autoclaving or chemical sterilization. Always clean your surgical instruments promptly after you use them. Proper cleaning lengthens the usable lifespan of surgical instruments.