Learn More
AdipoGen NIR 4d
SDP
Catalog No. 502560433
Encompass_Preferred
Chemical. CAS 162411-22-5. Formula C40H51ClN2O8S2. MW 787.4. Synthetic. Near-infrared near-IR, NIR fluorescent dye. Near-infrared near-IR, NIR dyes offer important advantages over traditional visible light dyes. Because cellular or tissue components produce minimal autofluorescence in the near-IR region, near-IR dyes have the potential to offer highly specific and sensitive detection in complex biological systems. Also, because light with wavelength in the near-IR region has strong tissue penetration, near-IR dyes are ideal for in vivo fluorescence imaging, an emerging field that has advanced rapidly in recent years. Max. absorbance 809nm
Catalog No. 50-256-0433 Supplier AdipoGen Supplier No. CDXN0035M050
Chemical. CAS 162411-22-5. Formula C40H51ClN2O8S2. MW 787.4. Synthetic. Near-infrared near-IR, NIR fluorescent dye. Near-infrared near-IR, NIR dyes offer important advantages over traditional visible light dyes. Because cellular or tissue components produce minimal autofluorescence in the near-IR region, near-IR dyes have the potential to offer highly specific and sensitive detection in complex biological systems. Also, because light with wavelength in the near-IR region has strong tissue penetration, near-IR dyes are ideal for in vivo fluorescence imaging, an emerging field that has advanced rapidly in recent years. Max. absorbance 809nm