Learn More
Medchemexpress LLC NIR-797-isothiocyanate | 152111-91-6 | MFCD01862952 | 99.3% | 880.14 g/mol | C45H50N3NaO6S4 | 10 MG
SDP
Catalog No. 5000297047
Encompass_Preferred
NIR-797-isothiocyanate is a near-infrared fluorescent dye bearing an isothiocyanate reactive group for covalent conjugation to primary amines. It is supplied as a brown to black solid intended for bioconjugation and imaging applications where near-infrared detection is required.
  • Near-infrared fluorescent dye suitable for imaging applications.
  • Isothiocyanate reactive group for covalent coupling to primary amines.
  • High purity (99.26%) for consistent labeling performance.
  • Molecular weight 880.14 g/mol aids reagent preparation and calculation.
  • Stable as a solid at -20°C; in solution store at -80°C for long-term.
Catalog No. 50-002-97047 Supplier Medchemexpress LLC Supplier No. HYD013410MG
NIR-797-isothiocyanate is a near-infrared fluorescent dye bearing an isothiocyanate reactive group for covalent conjugation to primary amines. It is supplied as a brown to black solid intended for bioconjugation and imaging applications where near-infrared detection is required.
  • Near-infrared fluorescent dye suitable for imaging applications.
  • Isothiocyanate reactive group for covalent coupling to primary amines.
  • High purity (99.26%) for consistent labeling performance.
  • Molecular weight 880.14 g/mol aids reagent preparation and calculation.
  • Stable as a solid at -20°C; in solution store at -80°C for long-term.