Learn More
AdipoGen NIR-797-isothiocyanate
SDP
Supplier:  AdipoGen CDXN0008M005
Encompass_Preferred
Chemical. CAS 152111-91-6. Formula C45H50N3NaO6S4. MW 880.14. Synthetic. The reagent, a cyanine dye, can be used as a label for the determination of proteins with NIR-fluorescent technology. The absorption-maxima of the reagent lies in a region of low interference by other biomolecules. The isothiocyanate-moiety is used to link the reagent to proteins which can sensitively be detected. Experiments to lower the detection-limits of immunoassays have been done.
Catalog No. 50-256-0444