Learn More
AdipoGen NIR-797-isothiocyanate,Chemical. CAS: 152111-91-6.
SDP
Supplier:  AdipoGen CDXN0008M025
Encompass_Preferred
NIR-797-isothiocyanate, AdipoGen Life Sciences. Chemical. CAS: 152111-91-6. Formula: C45H50N3NaO6S4. MW: 880.14. Synthetic. The reagent, a cyanine dye, can be used as a label for the determination of proteins with NIR-fluorescent technology. The absorption-maxima of the reagent lies in a region of low interference by other biomolecules. The isothiocyanate-moiety is used to link the reagent to proteins which can sensitively be detected. Experiments to lower the detection-limits of immunoassays have been done.
Catalog No. 50-595-819