Learn More
Sigma Aldrich NIR-II dye
Catalog No. 9008831MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
90-088-31MG 1 mg
1 options
Catalog No. 90-088-31MG Supplier Sigma Aldrich Supplier No. 9008831MG
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Stable solution in PBS. Miscible and stable in common buffers, serum and blood.Store in dark at 4°C.Suitable excitation wavelengths: 785 nm, 808 nm, Emission: 900-1400 nm, peak ∼1050 nm.Quantum yield (QY) ∼2% based on 0.5% QY of IR26 in 1,2-dichloroethane under 808 nm excitation. Quantum yield is ∼4X of common Hipco single walled carbon nanotubes.For in vivo imaging (e.g., vessels), inject ∼100-150 mug in ∼200 muL solution for a typical nude mouse but the dose can be varied.Circulation half time of the dye is about 1 hour.
Specifications
Quantity 1 mg
Synonym Near IR fluorescent dye