Learn More
Perkin Elmer US LLC NIR Integrating Sphere - InGaAs Detector
Catalog No. 502113286
Encompass_Preferred
Samples are placed directly onto the sample port or over the thin infrared transmitting window. You can also achieve diffuse transmission measurements by placing a sample at the slide mount beam input port. This accessory is compatible with the Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3286 Supplier Perkin Elmer US LLC Supplier No. L1272406
Add to Cart
Add to Cart
Request a Quote
Samples are placed directly onto the sample port or over the thin infrared transmitting window. You can also achieve diffuse transmission measurements by placing a sample at the slide mount beam input port. This accessory is compatible with the Spectrum 3/Frontier spectrometers.