Learn More
Perkin Elmer US LLC NIR Performance Validation Upgrade Kit
Catalog No. 502103692
Encompass_Preferred
The NIR Performance Validation (IPV) Upgrade Kit contains a replacement traceable reference material (TRM) to be used when the original TRM expires.
Catalog No. 50-210-3692 Supplier Perkin Elmer US LLC Supplier No. L1180490
The NIR Performance Validation (IPV) Upgrade Kit contains a replacement traceable reference material (TRM) to be used when the original TRM expires.