Learn More
Medchemexpress LLC HY-10619 50mg Medchemexpress, Niraparib CAS:1038915-60-4 Purity:>98%
SDP
Catalog No. 502028774
Encompass_Preferred
Medchemexpress, HY-10619 50mg Niraparib CAS:1038915-60-4 Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8774 Supplier Medchemexpress LLC Supplier No. HY1061950MG
Medchemexpress, HY-10619 50mg Niraparib CAS:1038915-60-4 Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.