Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niraparib carboxylic acid metabolite M1 | 1476777-06-6 | 99.7% | 321.37 g/mol | C19H19N3O2 | 10 MG
SDP

Catalog No. 5000297164
Encompass_Preferred

Niraparib metabolite M1 is the primary carboxylic acid metabolite of the PARP inhibitor niraparib, provided as a solid research reagent for in vitro and analytical use.

  • High purity (99.69%) suitable for analytical and research applications.
  • Molecular formula C19H19N3O2; molecular weight 321.37 g/mol.
  • Solid, light brown to brown appearance.
  • Supplied at 10 mg quantity.
  • Soluble in DMSO at ≥ 100 mg/mL (in vitro).
  • Recommended storage: powder at -20°C (up to 3 years); in solvent at -80°C (up to 2 years).
  • Intended for laboratory research use only; not for human therapeutic use.

Catalog No. 50-002-97164 Supplier Medchemexpress LLC Supplier No. HYG002310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Niraparib metabolite M1 is the primary carboxylic acid metabolite of the PARP inhibitor niraparib, provided as a solid research reagent for in vitro and analytical use.

  • High purity (99.69%) suitable for analytical and research applications.
  • Molecular formula C19H19N3O2; molecular weight 321.37 g/mol.
  • Solid, light brown to brown appearance.
  • Supplied at 10 mg quantity.
  • Soluble in DMSO at ≥ 100 mg/mL (in vitro).
  • Recommended storage: powder at -20°C (up to 3 years); in solvent at -80°C (up to 2 years).
  • Intended for laboratory research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.