Learn More
Medchemexpress LLC HY-10619A 5mg Medchemexpress, Niraparib hydrochloride CAS:1038915-64-8 Purity:>98%
SDP
Catalog No. 501968326
Encompass_Preferred
Medchemexpress, HY-10619A 5mg Niraparib HCl CAS:1038915-64-8 Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib hydrochloride (MK-4827 hydrochloride) leads to inhibition of repair of DNA damage and shows anti-tumor activity[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8326 Supplier Medchemexpress LLC Supplier No. HY10619A5MG
Medchemexpress, HY-10619A 5mg Niraparib HCl CAS:1038915-64-8 Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib hydrochloride (MK-4827 hydrochloride) leads to inhibition of repair of DNA damage and shows anti-tumor activity[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.