Learn More
Medchemexpress LLC Niraparib hydrochlor 10mg | 1038915-64-8 | 10MG
SDP
Catalog No. 5000244371
Encompass_Preferred
Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3 8 and 2 1 nM respectively Niraparib hydrochloride leads to inhibition of repair of DNA damage activates apoptosis and shows anti-tumor activity[1 [2 [3
Catalog No. 50-002-44371 Supplier Medchemexpress LLC Supplier No. HY10619A10MG
Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3 8 and 2 1 nM respectively Niraparib hydrochloride leads to inhibition of repair of DNA damage activates apoptosis and shows anti-tumor activity[1 [2 [3