Learn More
Medchemexpress LLC Niraparib (hydrochloride) | 1038915-64-8 | 99.4% | 356.85 g/mol | C19H21ClN4O | 50 MG
SDP
Catalog No. 5000243831
Encompass_Preferred
Niraparib hydrochloride is the hydrochloride salt of niraparib, a potent, orally bioavailable inhibitor of poly(ADP-ribose) polymerase 1 and 2 (PARP1/2). It inhibits DNA damage repair and induces apoptosis, making it useful for preclinical studies of DNA repair pathways and anti-tumor activity. Supplied as a solid 50 mg package for research use.
  • Potent PARP1 and PARP2 inhibition at nanomolar potency.
  • Effective tool for studying DNA damage repair and apoptosis.
  • Suitable for in vitro and in vivo preclinical assays.
  • Hydrochloride salt form improves handling and solubility.
  • Available as a 50 MG solid for small-scale experiments.
Catalog No. 50-002-43831 Supplier Medchemexpress LLC Supplier No. HY10619A50MG
Niraparib hydrochloride is the hydrochloride salt of niraparib, a potent, orally bioavailable inhibitor of poly(ADP-ribose) polymerase 1 and 2 (PARP1/2). It inhibits DNA damage repair and induces apoptosis, making it useful for preclinical studies of DNA repair pathways and anti-tumor activity. Supplied as a solid 50 mg package for research use.
  • Potent PARP1 and PARP2 inhibition at nanomolar potency.
  • Effective tool for studying DNA damage repair and apoptosis.
  • Suitable for in vitro and in vivo preclinical assays.
  • Hydrochloride salt form improves handling and solubility.
  • Available as a 50 MG solid for small-scale experiments.