Learn More
Medchemexpress LLC HY-G0023 5mg Medchemexpress, Niraparib metabolite M1 CAS:1476777-06-6 Purity:>98%
SDP
Catalog No. 501933247
Encompass_Preferred
Medchemexpress, HY-G0023 5mg Niraparib metabolite M1 CAS:1476777-06-6 Niraparib metabolite M1 is a metabolite of niraparib, and the latter one acts as a novel poly(ADP-Ribose) polymerase ( PARP ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3247 Supplier Medchemexpress LLC Supplier No. HYG00235MG
Medchemexpress, HY-G0023 5mg Niraparib metabolite M1 CAS:1476777-06-6 Niraparib metabolite M1 is a metabolite of niraparib, and the latter one acts as a novel poly(ADP-Ribose) polymerase ( PARP ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.