Learn More
Medchemexpress LLC HY-10619D 5mg Medchemexpress, Niraparib R-enantiomer CAS:1038915-58-0 Purity:>98%
SDP
Catalog No. 501968327
Encompass_Preferred
Medchemexpress, HY-10619D 5mg Niraparib R-enantiomer CAS:1038915-58-0 Niraparib R-enantiomer (MK-4827 R-enantiomer) is an excellent PARP1 inhibitor with IC50 of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8327 Supplier Medchemexpress LLC Supplier No. HY10619D5MG
Medchemexpress, HY-10619D 5mg Niraparib R-enantiomer CAS:1038915-58-0 Niraparib R-enantiomer (MK-4827 R-enantiomer) is an excellent PARP1 inhibitor with IC50 of 2.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.