Learn More
Medchemexpress LLC HY-10619B 50mg , Niraparib (tosylate) CAS:1038915-73-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10619B50MG
Encompass_Preferred
HY-10619B 50mg Niraparib (tosylate) CAS: 1038915-73-9 Niraparib tosylate (MK-4827 tosylate) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with an IC50 of 3.8 and 2.1 nM, respectively. Niraparib tosylate leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0481