Learn More
Selleck Chemical LLC Niraparib (MK-4827) tosylate 5mg 1038915-73-9 Niraparib tosylate, MK 4827 tosylate, ZEJULA
SDP
Supplier:  Selleck Chemical LLC S76255MG
Encompass_Preferred
Niraparib tosylate (MK-4827, ZEJULA) is a selective inhibitor of PARP1/PARP2 with IC50 of 3.8 nM/2.1 nM. Niraparib increases formation of PARP-DNA complexes resulting in DNA damage, apoptosis, and cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6362