Learn More
Selleck Chemical LLC Niraparib tosylate
SDP
Catalog No. 5000004046
Encompass_Preferred
Niraparib tosylate is a selective inhibitor of PARP1/PARP2 with IC50 of 3 8 nM/2 1 nM Niraparib increases formation of PARP-DNA complexes resulting in DNA damage apoptosis and cell death
Catalog No. 50-000-04046 Supplier Selleck Chemical LLC Supplier No. S762525MG
Add to Cart
Add to Cart
Niraparib tosylate is a selective inhibitor of PARP1/PARP2 with IC50 of 3 8 nM/2 1 nM Niraparib increases formation of PARP-DNA complexes resulting in DNA damage apoptosis and cell death