Learn More
TARGETMOL CHEMICALS INC Niraparib tosylate monohyrate
SDP
Catalog No. 502877209
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Niraparib, also know as MK-4827, is an inhibitor of poly (ADP-ribose) polymerase (PARP) with potential antineoplastic activity. MK4827 inhibits PARP activity, enhancing the accumulation of DNA strand breaks and promoting genomic instability and apoptosis. The PARP family of proteins detect and repair single strand DNA breaks by the base-excision repair (BER) pathway. Purity 99.09%
Catalog No. 50-287-7209 Supplier TARGETMOL CHEMICALS INC Supplier No. T949725MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Niraparib, also know as MK-4827, is an inhibitor of poly (ADP-ribose) polymerase (PARP) with potential antineoplastic activity. MK4827 inhibits PARP activity, enhancing the accumulation of DNA strand breaks and promoting genomic instability and apoptosis. The PARP family of proteins detect and repair single strand DNA breaks by the base-excision repair (BER) pathway. Purity 99.09%