Learn More
Selleck Chemical LLC Nirmatrelvir PF-07321332-5mg
SDP
Supplier:  Selleck Chemical LLC S98665MG
Encompass_Preferred
Nirmatrelvir (PF-07321332) is an reversible covalent inhibitor of SARS-CoV-2 main protease (Mpro, also referred to as 3CL protease) with an ki of 3.11 nM. PF-07321332 binds directly to the catalytic cysteine (Cys145) residue of the enzyme.
Catalog No. 50-238-9709