Learn More
Medchemexpress LLC HY-15185 10mg Medchemexpress, Nirogacestat CAS:1290543-63-3 Purity:>98%
SDP
Catalog No. 502029091
Encompass_Preferred
Medchemexpress, HY-15185 10mg Nirogacestat CAS:1290543-63-3 Nirogacestat (PF-3084014) is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor with an IC50 of 6.2 nM. Inhibition of Notch signaling by Nirogacestat while minimizing gastrointestinal toxicity presents a promising approach for research of Notch receptor-dependent cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9091 Supplier Medchemexpress LLC Supplier No. HY1518510MG
Medchemexpress, HY-15185 10mg Nirogacestat CAS:1290543-63-3 Nirogacestat (PF-3084014) is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor with an IC50 of 6.2 nM. Inhibition of Notch signaling by Nirogacestat while minimizing gastrointestinal toxicity presents a promising approach for research of Notch receptor-dependent cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.