Learn More
Selleck Chemical LLC Nirogacestat (PF-03084014) 5mg 1290543-63-3 PF-3084014
SDP
Supplier:  Selleck Chemical LLC S80185MG
Encompass_Preferred
Nirogacestat (PF-03084014, PF-3084014) is a selective gamma-secretase inhibitor with IC50 of 6.2 nM in a cell-free assay. Nirogacestat (PF-03084014, PF-3084014) induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8023