Learn More
Selleck Chemical LLC Nirogacestat
SDP
Catalog No. 5000003917
Encompass_Preferred
Nirogacestat (PF-03084014 PF-3084014) is a selective gamma-secretase inhibitor with IC50 of 6 2 nM in a cell-free assay Nirogacestat (PF-03084014 PF-3084014) induces apoptosis Phase 2
Catalog No. 50-000-03917 Supplier Selleck Chemical LLC Supplier No. S801825MG
Nirogacestat (PF-03084014 PF-3084014) is a selective gamma-secretase inhibitor with IC50 of 6 2 nM in a cell-free assay Nirogacestat (PF-03084014 PF-3084014) induces apoptosis Phase 2