Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nisin, G-Biosciences
SCI_ED

Catalog No. S157232
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157232 5 g
S139739 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157232 Supplier G-Biosciences Supplier No. RC1066
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nisin is a polypeptide antibiotic known as a lantibiotic. It has been isolated from Lactococcus lactis.

Produced by gram-positive bacteria, the role of nisin is to reduce competitive strain due to the presence of other gram-positive bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 3354.07

Specifications

Physical Form Powder
Quantity 5 g
Product Line Gibco
Boiling Point 2967°C
Percent Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.