Learn More
Medchemexpress LLC Nisin Z | 137061-46-2 | 3331.05 | 1 G
SDP
Catalog No. 5000398895
Encompass_Preferred
Nisin Z is an antimicrobial and anti-inflammatory peptide effective against Gram-positive bacteria and fungi, including C. albicans. This product is intended for research use only.
  • Antimicrobial and anti-inflammatory peptide
  • Effective against Gram-positive bacteria
  • Effective against fungi, such as C. albicans
  • Increases gingival fibroblasts detachment and differentiation
  • Decreases C. albicans adhesion to gingival cells
  • Inhibits germ tube formation by C. albicans
  • Reduces production of LPS-induced pro-inflammatory cytokines
  • Inhibits activation of ERK1/2 and p38 MAPK signaling pathway
  • Prevents pathological damage in mastitis mouse model
Catalog No. 50-003-98895 Supplier Medchemexpress LLC Supplier No. HYP34661G
Nisin Z is an antimicrobial and anti-inflammatory peptide effective against Gram-positive bacteria and fungi, including C. albicans. This product is intended for research use only.
  • Antimicrobial and anti-inflammatory peptide
  • Effective against Gram-positive bacteria
  • Effective against fungi, such as C. albicans
  • Increases gingival fibroblasts detachment and differentiation
  • Decreases C. albicans adhesion to gingival cells
  • Inhibits germ tube formation by C. albicans
  • Reduces production of LPS-induced pro-inflammatory cytokines
  • Inhibits activation of ERK1/2 and p38 MAPK signaling pathway
  • Prevents pathological damage in mastitis mouse model