Learn More
Medchemexpress LLC Nisin Z | 137061-46-2 | 97.1% | 3331.05 | 5 G
SDP
Catalog No. 5000398899
Encompass_Preferred
Nisin Z is an antimicrobial and anti-inflammatory peptide effective against Gram-positive bacteria and fungi, including C. albicans. It demonstrates a range of biological activities in both in vitro and in vivo models.
  • Increases gingival fibroblasts detachment and differentiation.
  • Decreases C. albicans adhesion to gingival cells.
  • Inhibits germ tube formation by C. albicans.
  • Reduces production of LPS-induced pro-inflammatory cytokines.
  • Inhibits activation of ERK1/2 and p38 MAPK signaling pathway.
  • Reduces C. albicans growth in cell viability assays.
  • Inhibits phosphorylation of p38 and ERK1/2.
  • Prevents pathological damage in mastitis mouse models.
  • Reduces inflammatory cell infiltration and cytokine levels.
Catalog No. 50-003-98899 Supplier Medchemexpress LLC Supplier No. HYP34665G
Nisin Z is an antimicrobial and anti-inflammatory peptide effective against Gram-positive bacteria and fungi, including C. albicans. It demonstrates a range of biological activities in both in vitro and in vivo models.
  • Increases gingival fibroblasts detachment and differentiation.
  • Decreases C. albicans adhesion to gingival cells.
  • Inhibits germ tube formation by C. albicans.
  • Reduces production of LPS-induced pro-inflammatory cytokines.
  • Inhibits activation of ERK1/2 and p38 MAPK signaling pathway.
  • Reduces C. albicans growth in cell viability assays.
  • Inhibits phosphorylation of p38 and ERK1/2.
  • Prevents pathological damage in mastitis mouse models.
  • Reduces inflammatory cell infiltration and cytokine levels.