Learn More
Cayman Chemical Nisoldipine, Purity: ≥97%, CAS: 63675-72-9, 100 mg
SDP
Catalog No. 502982182
Encompass_Preferred
A calcium channel inhibitor; binds to calcium channels in isolated rat ventricular membranes (Kd = 0.04 nM) and inhibits calcium uptake by smooth muscle cells; reduces ventricular tachycardia and fibrillization and increases survival in a rat model of ventricular arrhythmias induced by myocardial ischemia at 3 mg/kg; dietary administration reduces reduces systolic blood pressure in spontaneously hypertensive rats at 50 and 100 mg/kg
Catalog No. 50-298-2182 Supplier Cayman Chemical Supplier No. 20998100 MG
A calcium channel inhibitor; binds to calcium channels in isolated rat ventricular membranes (Kd = 0.04 nM) and inhibits calcium uptake by smooth muscle cells; reduces ventricular tachycardia and fibrillization and increases survival in a rat model of ventricular arrhythmias induced by myocardial ischemia at 3 mg/kg; dietary administration reduces reduces systolic blood pressure in spontaneously hypertensive rats at 50 and 100 mg/kg