Learn More
Medchemexpress LLC Nisoxetine | 53179-07-0 | 98.6% | 100 MG
SDP
Catalog No. 5000413468
Encompass_Preferred
Nisoxetine is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It functions as an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels. This product is intended for research use only.
  • Potently and selectively inhibits the noradrenaline transporter (NET) with a Kd of 0.76 nM.
  • Acts as both an antidepressant and a local anesthetic.
  • Can block voltage-gated sodium channels.
  • Available in multiple quantities.
  • Comes with a data sheet, COA, SDS, and handling instructions.
  • Detailed solubility information provided for in vitro use.
Catalog No. 50-004-13468 Supplier Medchemexpress LLC Supplier No. HYB1704100MG
Nisoxetine is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It functions as an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels. This product is intended for research use only.
  • Potently and selectively inhibits the noradrenaline transporter (NET) with a Kd of 0.76 nM.
  • Acts as both an antidepressant and a local anesthetic.
  • Can block voltage-gated sodium channels.
  • Available in multiple quantities.
  • Comes with a data sheet, COA, SDS, and handling instructions.
  • Detailed solubility information provided for in vitro use.