Learn More
Medchemexpress LLC Nisoxetine | 53179-07-0 | MFCD00865443 | 98.6% | 271.35 | C17H21NO2 | 10 MG
SDP
Catalog No. 5000298959
Encompass_Preferred
Nisoxetine is a selective norepinephrine transporter (NET) inhibitor intended for research use. It is supplied as a high-purity solid suitable for pharmacology, binding, and analytical studies.
  • Selective norepinephrine transporter inhibitor for research use only.
  • High purity (≈98.6%) suitable for analytical assays.
  • Molecular weight 271.35 and formula C17H21NO2.
  • Available in sizes from 5-500 mg to support assay scale-up.
  • Supplied with analytical documentation (COA, LCMS, NMR, SDS).
Catalog No. 50-002-98959 Supplier Medchemexpress LLC Supplier No. HYB170410MG
Nisoxetine is a selective norepinephrine transporter (NET) inhibitor intended for research use. It is supplied as a high-purity solid suitable for pharmacology, binding, and analytical studies.
  • Selective norepinephrine transporter inhibitor for research use only.
  • High purity (≈98.6%) suitable for analytical assays.
  • Molecular weight 271.35 and formula C17H21NO2.
  • Available in sizes from 5-500 mg to support assay scale-up.
  • Supplied with analytical documentation (COA, LCMS, NMR, SDS).