Learn More
Medchemexpress LLC Nisoxetine hydrochloride | 57754-86-6 | 99.9% | 25 MG
SDP
Catalog No. 5000414184
Encompass_Preferred
Nisoxetine hydrochloride is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It is an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Functions as an antidepressant
  • Acts as a local anesthetic
  • Blocks voltage-gated sodium channels
Catalog No. 50-004-14184 Supplier Medchemexpress LLC Supplier No. 25MGHYB1704A
Nisoxetine hydrochloride is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It is an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Functions as an antidepressant
  • Acts as a local anesthetic
  • Blocks voltage-gated sodium channels