Learn More
Medchemexpress LLC Nisoxetine hydrochloride | 57754-86-6 | MFCD00078426 | 99.92% | 5 MG
SDP
Catalog No. 5000412755
Encompass_Preferred
Nisoxetine hydrochloride is a potent and selective inhibitor of the noradrenaline transporter (NET), with a Kd of 0.76 nM. It functions as an antidepressant and local anesthetic by blocking voltage-gated sodium channels.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Acts as an antidepressant
  • Functions as a local anesthetic
  • Blocks voltage-gated sodium channels
Catalog No. 50-004-12755 Supplier Medchemexpress LLC Supplier No. HYB1704A5MG
Nisoxetine hydrochloride is a potent and selective inhibitor of the noradrenaline transporter (NET), with a Kd of 0.76 nM. It functions as an antidepressant and local anesthetic by blocking voltage-gated sodium channels.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Acts as an antidepressant
  • Functions as a local anesthetic
  • Blocks voltage-gated sodium channels