Learn More
Ihc World LLC NISSLSTAIN KIT
SDP
Catalog No. NC0098757
Encompass_Preferred
This kit is used for the detection of Nissl body in the cytoplasm of neurons on formalin-fixed, paraffin embedded tissue sections, as well as frozen sections. The Nissl body will be stained purple-blue. This stain is commonly used for identifying the
Catalog No. NC0098757 Supplier Ihc World LLC Supplier No. IW3007
This kit is used for the detection of Nissl body in the cytoplasm of neurons on formalin-fixed, paraffin embedded tissue sections, as well as frozen sections. The Nissl body will be stained purple-blue. This stain is commonly used for identifying the