Learn More
METTLER TOLEDO 3P INDUSTRIAL NIST/DIN buffer pH 6.865, 250mL
Catalog No. NC1721021
Encompass_Preferred
These buffers are specified to three decimal places and thus offer the highest possible precision.
Catalog No. NC1721021 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51350054
These buffers are specified to three decimal places and thus offer the highest possible precision.