Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO 3P INDUSTRIAL NIST Buffer pH 9.180, 250 mL bottle, NIST/DIN certified

Catalog No. NC2211085
Encompass_Preferred

The METTLER TOLEDO 51350056 is a high-precision buffer solution tailored for laboratory pH meter calibration. It is NIST/DIN certified, ensuring accuracy within scientific measurements. This solution is designed to deliver precise readings, traceable to NIST primary standards, and is available in a convenient 250 mL bottle. A quality inspection certificate is included for documentation, enhancing its reliability for calibration tasks.

  • pH Value at 25 °C: 9.180
  • Volume: 250 mL
  • Certification: NIST/DIN certified
  • Traceability: To NIST primary standards
  • Precision: Three decimal places
  • Uncertainty: ±0.015 pH
  • Shelf Life: 12 months
  • Packaging Type: Bottle
  • Application: pH meter calibration
  • Country of Origin: Switzerland
  • Documentation: Quality inspection certificate included, SDS available online

This item is not returnable. View return policy
Catalog No. NC2211085 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51350056
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The METTLER TOLEDO 51350056 is a high-precision buffer solution tailored for laboratory pH meter calibration. It is NIST/DIN certified, ensuring accuracy within scientific measurements. This solution is designed to deliver precise readings, traceable to NIST primary standards, and is available in a convenient 250 mL bottle. A quality inspection certificate is included for documentation, enhancing its reliability for calibration tasks.

  • pH Value at 25 °C: 9.180
  • Volume: 250 mL
  • Certification: NIST/DIN certified
  • Traceability: To NIST primary standards
  • Precision: Three decimal places
  • Uncertainty: ±0.015 pH
  • Shelf Life: 12 months
  • Packaging Type: Bottle
  • Application: pH meter calibration
  • Country of Origin: Switzerland
  • Documentation: Quality inspection certificate included, SDS available online

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.