Learn More
Cole-Parmer InnoCal NIST-Traceable Calibration; Refractometer
Supplier:  Cole-Parmer 1710500
Encompass
This NIST-traceable calibrations is for multimeters, calibrators, oscilloscopes, and more. It provides traceability to NIST and lists the environmental conditions present at time of calibration. Its certificate lists actual values, uncertainties, and tolerances. Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards. It does not have an accredited certificate.
Catalog No. NC0734904