Learn More
Description
- It ensures accurate, standardized pH meter calibration, ideal for acid-range applications in labs and industries.
- The size supports multiple calibrations, helping maintain traceability, measurement confidence, and regulatory compliance.
Specifications
Specifications
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 3.776 |
| Physical Form | Solution |
| Quantity | 250 mL |
| Solution Type | pH Buffer |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.