Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
GREENER_CHOICE

Catalog No. 01914332 Shop All METTLER TOLEDO Products
Change view
Click to view available options
pH Range:
10.012 (at 25°C)
4.006 (at 25°C)
6.865 (at 25°C)
9.180 (at 25°C)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
01-914-332 9.180 (at 25°C)
01-914-330 4.006 (at 25°C)
01-914-331 6.865 (at 25°C)
01-914-333 10.012 (at 25°C)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-914-332 Supplier Mettler Toledo™ Supplier No. 30111138
Add to Cart
Add to Cart

Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.

pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)

  • NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
  • Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
  • A test certificate and detailed label with a temperature table comes with every box
  • All SDS and labels contain information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY

Specifications

Description NIST/DIN Buffer pH 9.180 sachets 30x20mL
For Use With (Equipment) pH Electrode
For Use With (Application) pH measurement, electrode calibration
NIST Traceable Yes
Packaging Sachet
pH Range 9.180 (at 25°C)
Quantity 30 x 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.