Learn More
Thermco Products Inc NIST Pocket Thermometer w/Probe Digital Thermometer
Catalog No. NC1692837
Encompass_Preferred
NIST Traceable w/Certificate. Pocket Size 4.5” x 2.5” x .5”. Attached Bench Stand. Range: -50º to 300ºC (-58° to 572°F).
Catalog No. NC1692837 Supplier Thermco Products Inc Supplier No. ACC550DIG
NIST Traceable w/Certificate. Pocket Size 4.5” x 2.5” x .5”. Attached Bench Stand. Range: -50º to 300ºC (-58° to 572°F).